Learn More
Apexbio Technology LLC Biotin-HPDP, 100mg. Cas: 129179-83-5 MFCD: MFCD00078541. Sulfhydryl-reactive biotinylation reagent, pyridyldithiol-activated
SDP
Supplier:  Apexbio Technology LLC A8008100
Encompass_Preferred
Pyridyldithiol-biotin compound, reacts with thiol groups on cysteine residues and forms reversible disulfide bonds 129179-83-5. MFCD00078541
Catalog No. 50-101-0614