Learn More
Cayman Chemical BIotIn-HPDP 10mg
SDP
Catalog No. 502971347
Encompass_Preferred
A sulfhydryl-reactive biotinylation reagent that forms a reversible disulfide linkage, used to label protein cysteines and other substrates that contain sulfhydryl groups; EAsily linked with target substrates at pH 6.5 to 7.5 in buffers such as PBS; disulfide linkage can be cleaved by reducing agents
Catalog No. 50-297-1347 Supplier Cayman Chemical Supplier No. 1645910 MG
A sulfhydryl-reactive biotinylation reagent that forms a reversible disulfide linkage, used to label protein cysteines and other substrates that contain sulfhydryl groups; EAsily linked with target substrates at pH 6.5 to 7.5 in buffers such as PBS; disulfide linkage can be cleaved by reducing agents