Learn More
Medchemexpress LLC Biotin-HPDP | 129179-83-5 | MFCD00078541 | 99.7% | 539.78 g/mol | C24H37N5O3S3 | 5 MG
SDP
Catalog No. 5000222797
Encompass_Preferred
Biotin-HPDP is a sulfhydryl-reactive biotinylation reagent used to label and affinity-purify cysteine-containing proteins. It forms a reversible disulfide linkage to thiol groups, enabling reversible labeling and recovery of native proteins. The compound has a molecular weight of 539.78 g/mol and a reported purity of 99.73%.
  • Sulfhydryl-reactive biotinylation reagent that targets free thiols.
  • Forms reversible disulfide linkage enabling recovery of native proteins.
  • Suitable for labeling and affinity purification of cysteine-containing proteins.
  • High reported purity for reproducible experimental results.
  • Soluble in DMSO and compatible with common in vivo formulations.
  • Available in small research sizes for laboratory use.
Catalog No. 50-002-22797 Supplier Medchemexpress LLC Supplier No. HY1367695MG
Biotin-HPDP is a sulfhydryl-reactive biotinylation reagent used to label and affinity-purify cysteine-containing proteins. It forms a reversible disulfide linkage to thiol groups, enabling reversible labeling and recovery of native proteins. The compound has a molecular weight of 539.78 g/mol and a reported purity of 99.73%.
  • Sulfhydryl-reactive biotinylation reagent that targets free thiols.
  • Forms reversible disulfide linkage enabling recovery of native proteins.
  • Suitable for labeling and affinity purification of cysteine-containing proteins.
  • High reported purity for reproducible experimental results.
  • Soluble in DMSO and compatible with common in vivo formulations.
  • Available in small research sizes for laboratory use.