Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Biotin-HRP (biotinylated peroxidase)

Catalog No. 432040
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-204-0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-204-0 Supplier Invitrogen™ Supplier No. 432040
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-HRP (biotinylated peroxidase)

Reactivity: Reacts with streptavidin and avidin conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator at 2°C to 8°C
Form Liquid
Label or Dye HRP (Horseradish Peroxidase)
Shipping Condition Wet Ice
Quantity 5 mg

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.