Learn More
Bon Opus Biosciences Biotinylated Human 5T4 (C-Avi-6His) -20ug
SDP
Supplier:  Bon Opus Biosciences CY2920UG
Encompass_Preferred
Biotinylated Recombinant Human Trophoblast Glycoprotein is produced by our Mammalian expression system and the target gene encoding Ser32-Ser355 is expressed with a Avi, 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q13641. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# CY29
Catalog No. 50-208-0522