Learn More
Bon Opus Biosciences Biotinylated Human BCMA (C-Fc-Avi) -20ug
SDP
Supplier:  Bon Opus Biosciences CY2220UG
Encompass_Preferred
Biotinylated Recombinant Human B-cell Maturation Protein is produced by our Mammalian expression system and the target gene encoding Met1-Ala54 is expressed with a Fc, Avi tag at the C-terminus. Expression system: Human Cells. Accession# Q02223. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# CY22
Catalog No. 50-208-0514