Learn More
Bon Opus Biosciences Biotinylated Human CD30 (C-6His-Avi) -100ug
SDP
Catalog No. 502080483
Encompass_Preferred
Biotinylated Recombinant Human Epithelial Cell Adhesion Molecule is produced by our Mammalianexpression system and the target gene encoding Gln24-Lys265(Met115Thr) is expressed with a 6His, Avitag at the C-terminus. Expression system: Human Cells. Accession# P28908. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mMNaCl, pH7.4. Bon Opus Cat# CY04
Catalog No. 50-208-0483 Supplier Bon Opus Biosciences Supplier No. CY04100UG
Add to Cart
Add to Cart
Biotinylated Recombinant Human Epithelial Cell Adhesion Molecule is produced by our Mammalianexpression system and the target gene encoding Gln24-Lys265(Met115Thr) is expressed with a 6His, Avitag at the C-terminus. Expression system: Human Cells. Accession# P28908. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mMNaCl, pH7.4. Bon Opus Cat# CY04