Learn More
Bon Opus Biosciences Biotinylated Human CD30 (C-6His-Avi) -20ug
SDP
Catalog No. 502080482
Encompass_Preferred
Biotinylated Recombinant Human Epithelial Cell Adhesion Molecule is produced by our Mammalianexpression system and the target gene encoding Gln24-Lys265(Met115Thr) is expressed with a 6His, Avitag at the C-terminus. Expression system: Human Cells. Accession# P28908. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mMNaCl, pH7.4. Bon Opus Cat# CY04
Catalog No. 50-208-0482 Supplier Bon Opus Biosciences Supplier No. CY0420UG
Add to Cart
Add to Cart
Biotinylated Recombinant Human Epithelial Cell Adhesion Molecule is produced by our Mammalianexpression system and the target gene encoding Gln24-Lys265(Met115Thr) is expressed with a 6His, Avitag at the C-terminus. Expression system: Human Cells. Accession# P28908. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mMNaCl, pH7.4. Bon Opus Cat# CY04