Learn More
Bon Opus Biosciences Biotinylated Human CEACAM5 (C-6His-Avi) -100ug
SDP
Supplier:  Bon Opus Biosciences CY23100UG
Encompass_Preferred
Biotinylated Recombinant Human Carcinoembryonic Antigen-Related Cell Adhesion Molecule 5 is produced by our Mammalian expression system and the target gene encoding Lys35-Ala685 is expressed with a Avi, 6His tag at the C-terminus. Expression system: Human Cells. Accession# P06731. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# CY23
Catalog No. 50-208-0517