Learn More
Bon Opus Biosciences Biotinylated Human FGL1 (C-Avi-6His) -100ug
SDP
Supplier:  Bon Opus Biosciences CY21100UG
Encompass_Preferred
Recombinant Human Fibrinogen-like Protein 1 is produced by our Mammalian expression system and the target gene encoding Leu23-Ile312 is expressed with a Avi, 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q08830. Formulation: Supplied as a 0.2 μm filtered solution of 20mM Tris, 300mM Nacl, 10%Glycerol, 2mM EDTA, pH8.0. Bon Opus Cat# CY21
Catalog No. 50-208-0513