Learn More
Bon Opus Biosciences Biotinylated Human FOLR1 (C-6His-Avi) -100ug
SDP
Supplier:  Bon Opus Biosciences CY16100UG
Encompass_Preferred
Biotinylated Recombinant Human Folate Receptor alpha is produced by our Mammalian expression system and the target gene encoding Arg25-Ser234 is expressed with a 6His, Avi tag at the C-terminus.  Expression system: Human Cells. Accession# P15328. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH7.4. Bon Opus Cat# CY16
Catalog No. 50-208-0505