Learn More
Bon Opus Biosciences Biotinylated Human GPC3 (C-6His-Avi) -20ug
SDP
Supplier:  Bon Opus Biosciences CY1520UG
Encompass_Preferred
Biotinylated Recombinant Human Glypican-3 is produced by our Mammalian expression system and the target gene encoding Gln25-His559 is expressed with a 6His, Avi tag at the C-terminus. Expression system: Human Cells. Accession# P51654. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# CY15
Catalog No. 50-208-0502