Learn More
Bon Opus Biosciences Biotinylated Human IL-3RA (C-Fc-Avi) -100ug
SDP
Supplier:  Bon Opus Biosciences CY01100UG
Encompass_Preferred
Biotinylated Recombinant Human Interleukin-3 Receptor Subunit Alpha is produced by our Mammalian expression system and the target gene encoding Thr19-Arg305 is expressed with a Fc, Avi tag at the C-terminus. Expression system: Human Cells. Accession# P26951. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# CY01
Catalog No. 50-208-0478