Learn More
Bon Opus Biosciences Biotinylated Recombinant Human MerTK (C-hFc-Avi) 25ug
SDP
Supplier:  Bon Opus Biosciences BP111625UG
Encompass_Preferred
Biotinylated Recombinant Human MerTK (C-hFc-Avi) is produced by Human Cells. The target gene encoding Arg26-Ala499 is expressed with a C-hFc-Avi tag. Accession# Q12866-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1116
Catalog No. 50-217-2711