Learn More
Bon Opus Biosciences Biotinylated Human PD-1 (C-6His-Avi) -100ug
SDP
Supplier:  Bon Opus Biosciences CY18100UG
Encompass_Preferred
Biotinylated Recombinant Human Programmed Cell Death Protein 1 is produced by our Mammalian expression system and the target gene encoding Pro21-Gln167 is expressed with a 6His, Avi tag at the C-terminus. Expression system: Human Cells. Accession# Q15116. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# CY18
Catalog No. 50-208-0507