Learn More
Bon Opus Biosciences Biotinylated Human ROR1 (C-6His-Avi) -100ug
SDP
Supplier:  Bon Opus Biosciences CY06100UG
Encompass_Preferred
Biotinylated Recombinant Human Inactive Tyrosine-protein Kinase Transmembrane Receptor ROR1 isproduced by our Mammalian expression system and the target gene encoding Gln30-Glu403 is expressedwith a 6His, Avi tag at the C-terminus. Expression system: Human Cells. Accession# Q01973. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# CY06
Catalog No. 50-208-0487