Learn More
Bon Opus Biosciences Biotinylated Human Siglec-15 (C-Fc-Avi) -100ug
SDP
Supplier:  Bon Opus Biosciences CY14100UG
Encompass_Preferred
Biotinylated Recombinant Human Sialic Acid-binding Ig-like Lectin 15 is produced by our Mammalian expression system and the target gene encoding Phe20-Thr263 is expressed with a Fc, Avi tag at the C-terminus. Expression system: Human Cells. Accession# Q6ZMC9. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, 150mM NaCl, 0.3% Chaps, 5% Trehalose, pH 7.4. Bon Opus Cat# CY14
Catalog No. 50-208-0501