Learn More
Bon Opus Biosciences Biotinylated Human SIRPA (C-6His-Avi) -20ug
SDP
Supplier:  Bon Opus Biosciences CY2820UG
Encompass_Preferred
Biotinylated Recombinant Human Signal-Regulatory Protein Alpha 1 is produced by our Mammalian expression system and the target gene encoding Glu31-Arg370 is expressed with a 6His, Avi tag at the C-terminus. Expression system: Human Cells. Accession# P78324. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# CY28
Catalog No. 50-208-0520