Learn More
Bon Opus Biosciences Biotinylated Human TIGIT (C-6His-Avi) -20ug
SDP
Supplier:  Bon Opus Biosciences CY1320UG
Encompass_Preferred
Biotinylated Recombinant Human T cell Immunoreceptor with Ig and ITIM domains is produced by our Mammalian expression system and the target gene encoding Met22-Pro141 is expressed with a 6His, Avi tag at the C-terminus. Expression system: Human Cells. Accession# Q495A1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# CY13
Catalog No. 50-208-0498