Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human B7-H4 Protein (recommended for biopanning), Avi Tag (Avitag™);HEK293;25UG;B74,B7-H4,VTCN1,B7H4,B7S1,B7h.5,VTCN-1,B7-S1
Supplier: ACROBiosystems B74H82E225UG
Recombinant Protein;Biotinylated Human B7-H4 Protein (recommended for biopanning), Avi Tag (Avitag™);25UG;Biotinylated Human B7-H4, His Tag (B74-H82E2) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Ala 258 (Accession # NP_078902).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 28 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-B7-H4 mAb,Human IgG1 at 2 μg/mL (100 μL/well) can bind Biotinylated Human B7-H4, His Tag (Cat. No. B74-H82E2) with a linear range of 3-48 ng/mL (QC tested).