Learn More
ACROBiosystems BIOTIN HUM CD155, FC TAG 25UG
Supplier:  ACROBiosystems CD5H82F625UG
Encompass_Preferred
Biotinylated Human CD155, Fc Tag (CD5-H82F6) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accession # AAH15542).
Catalog No. 50-113-8440