Learn More
ACROBiosystems BIOTIN HUM CD19 FC 200UG
Catalog No. 501017458
Encompass_Preferred
MABSol Biotinylated Human CD19, Fc Tag (CD9-H8259) is expressed from human HEK293 cells. It contains AA Pro 20 - Lys 291 (Accession # AAH06338). It is the biotinylated form of Human CD19 Protein, Fc Tag (Cat # CD9-H5259).
Catalog No. 50-101-7458 Supplier ACROBiosystems Supplier No. CD9H8259200UG
MABSol Biotinylated Human CD19, Fc Tag (CD9-H8259) is expressed from human HEK293 cells. It contains AA Pro 20 - Lys 291 (Accession # AAH06338). It is the biotinylated form of Human CD19 Protein, Fc Tag (Cat # CD9-H5259).