Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Siglec-3 / CD33 Protein, Avi Tag (Avitag™);HEK293;25UG;CD3,CD33,SIGLEC3,SIGLEC-3,gp67
Catalog No. 501138404
Encompass_Preferred
Recombinant Protein;Biotinylated Human Siglec-3 / CD33 Protein, Avi Tag (Avitag™);25UG;Biotinylated Human Siglec-3, His Tag (CD3-H82E7) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 29.4 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized anti human CD33 MAb, Mouse IgG1 (Clone # 6C5/2) at 2 μg/mL (100 μL/well) can bind Biotinylated Human Siglec-3, His Tag (Cat. No. CD3-H82E7) with a linear range of 0.078-2.5 ng/mL (QC tested).
Catalog No. 50-113-8404 Supplier ACROBiosystems Supplier No. CD3H82E725UG
Recombinant Protein;Biotinylated Human Siglec-3 / CD33 Protein, Avi Tag (Avitag™);25UG;Biotinylated Human Siglec-3, His Tag (CD3-H82E7) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 29.4 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized anti human CD33 MAb, Mouse IgG1 (Clone # 6C5/2) at 2 μg/mL (100 μL/well) can bind Biotinylated Human Siglec-3, His Tag (Cat. No. CD3-H82E7) with a linear range of 0.078-2.5 ng/mL (QC tested).