Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CTLA-4, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;CT4,CTLA4,CD152,CELIAC3,CTLA-4,GRD4,GSE,ICOS,IDDM12
Catalog No. 501017602
Encompass_Preferred
Recombinant Protein;Biotinylated Human CTLA-4, Fc Tag, Avi Tag (Avitag™);25UG;MABSol Biotinylated Human CTLA-4, Fc Tag (CT4-H82F3) is expressed from human HEK293 cells. It contains AA Ala 37 - Phe 162 (Accession # NP_005205.2).;This protein carries a human IgG1 Fc fragment at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 42 kDa. The protein migrates as 50-55 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Human B7-2, Fc Tag (Cat. No. CD6-H5257) at 5 g/mL (100 l/well),can bind Biotinylated Human CTLA-4, Fc tag (Cat. No. CT4-H82F3) with a linear range of 2-30 ng/mL (QC tested).;Immobilized Human B7-1, Fc Tag (Cat. No. B71-H5259) at 5 g/mL (100 l/well),can bind Biotinylated Human CTLA-4, Fc tag (Cat. No. CT4-H82F3) with a linear range of 0.1-8 ng/mL (Routinely tested).
Catalog No. 50-101-7602 Supplier ACROBiosystems Supplier No. CT4H82F325UG
Recombinant Protein;Biotinylated Human CTLA-4, Fc Tag, Avi Tag (Avitag™);25UG;MABSol Biotinylated Human CTLA-4, Fc Tag (CT4-H82F3) is expressed from human HEK293 cells. It contains AA Ala 37 - Phe 162 (Accession # NP_005205.2).;This protein carries a human IgG1 Fc fragment at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 42 kDa. The protein migrates as 50-55 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Human B7-2, Fc Tag (Cat. No. CD6-H5257) at 5 g/mL (100 l/well),can bind Biotinylated Human CTLA-4, Fc tag (Cat. No. CT4-H82F3) with a linear range of 2-30 ng/mL (QC tested).;Immobilized Human B7-1, Fc Tag (Cat. No. B71-H5259) at 5 g/mL (100 l/well),can bind Biotinylated Human CTLA-4, Fc tag (Cat. No. CT4-H82F3) with a linear range of 0.1-8 ng/mL (Routinely tested).