Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human DNAM-1 / CD226 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;DN1,DNAM1,CD226,CD226 antigen,DNAM-1,PTA1
Supplier: ACROBiosystems DN1H82F925UG
Recombinant Protein;Biotinylated Human DNAM-1 / CD226 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human DNAM-1, Fc Tag (DN1-H82F9) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Accession # NP_006557).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 54.4 kDa. The protein migrates as 66-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rhesus macaque CD155 (Cat. No. CD5-R52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human DNAM-1, Fc Tag (Cat. No. DN1-H82F9) with a linear range of 10-156 ng/mL (QC tested).;FACS assay shows Biotinylated Human DNAM-1 Protein, Fc Tag, Avi Tag (Cat. No. DN1-H82F9) can bind to 293T cell overexpressing human CD155. The concentration of DNAM-1 is 0.03 μg/ml (Routinely tested).