Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human EGFRvIII, Avi Tag (Avitag™);HEK293;200UG;EGR,EGFRvIII
Catalog No. 501138397
Encompass_Preferred
Recombinant Protein;Biotinylated Human EGFRvIII, Avi Tag (Avitag™);200UG;MABSolBiotinylated Human EGFRvIII (Cat. No. EGR-H82E0) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (30-297: Val Cys Gln...Cys Pro Arg Gly) (Accession # NP_005219.2). ;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 41.3 kDa. As a result of glycosylation, the protein migrates as 60-70 kDa under reducing (R) condition, and 60-70 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized CetuxiMabat 2 g/mL (100 L/well) can bind Biotinylated Human EGFRvIII (Cat. No. EGR-H82E0) with a linear range of 0.4-25 ng/mL (QC tested).
Catalog No. 50-113-8397 Supplier ACROBiosystems Supplier No. EGRH82E0200UG
Recombinant Protein;Biotinylated Human EGFRvIII, Avi Tag (Avitag™);200UG;MABSolBiotinylated Human EGFRvIII (Cat. No. EGR-H82E0) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (30-297: Val Cys Gln...Cys Pro Arg Gly) (Accession # NP_005219.2). ;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 41.3 kDa. As a result of glycosylation, the protein migrates as 60-70 kDa under reducing (R) condition, and 60-70 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized CetuxiMabat 2 g/mL (100 L/well) can bind Biotinylated Human EGFRvIII (Cat. No. EGR-H82E0) with a linear range of 0.4-25 ng/mL (QC tested).