Learn More
ACROBiosystems BIOTIN HUM FCGRT / B2M 200UG
Catalog No. 501017795
Encompass_Preferred
MABSol Biotinylated Human FcRn / FCGRT & B2M Heterodimer Protein (FCM-H82W4) is expressed from human HEK293 cells. It contains AA Ala 24 - Ser 297 (FCGRT) & Ile 21 - Met 119 (B2M) (Accession # NP_001129491.1 (FCGRT) & AAH32589 (B2M)).
Catalog No. 50-101-7795 Supplier ACROBiosystems Supplier No. FCMH82W4200UG
MABSol Biotinylated Human FcRn / FCGRT & B2M Heterodimer Protein (FCM-H82W4) is expressed from human HEK293 cells. It contains AA Ala 24 - Ser 297 (FCGRT) & Ile 21 - Met 119 (B2M) (Accession # NP_001129491.1 (FCGRT) & AAH32589 (B2M)).