Learn More
ACROBiosystems BIOTIN HUM IL-23A&IL-12B 25UG
Catalog No. 501138461
Encompass_Preferred
Biotinylated Human IL-23 alpha & IL-12 beta Heterodimer (ILB-H82W6) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Pro 189 (IL23A) & Ile 23 - Ser 328 (IL12B) (Accession # NP_057668 (IL23A) & AAH67499 (IL12B)).
Catalog No. 50-113-8461 Supplier ACROBiosystems Supplier No. ILBH82W625UG
Biotinylated Human IL-23 alpha & IL-12 beta Heterodimer (ILB-H82W6) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Pro 189 (IL23A) & Ile 23 - Ser 328 (IL12B) (Accession # NP_057668 (IL23A) & AAH67499 (IL12B)).