Learn More
ACROBiosystems BIOTIN HUM VEGF165, HIS 25UG
Catalog No. 501138457
Encompass_Preferred
Biotinylated Human VEGF165, His Tag (VE5-H82Q0) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Accession # P15692-4).
Catalog No. 50-113-8457 Supplier ACROBiosystems Supplier No. VE5H82Q025UG
Biotinylated Human VEGF165, His Tag (VE5-H82Q0) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Accession # P15692-4).