Learn More
AnaSpec BIOTIN-HUMAN AMYLIN PEPTIDE
Supplier:  AnaSpec AS6445105
Encompass
Biotin-Amylin (1-37), amide, human[Biotin-KCNTATCATQRLANFLVHSSNNFGAILSSTNVGSNTY-NH2 (disulfide bridge: 2-7)] ; MW 4129.7; (0.5 mg) This amidated amylin (1-37) peptide has a biotin conjugated on the N-terminus. Amylin (1-37) or IAPP (Islet Amyloid Polypeptide Precursor) is a major constituent of protein deposits identified in the Islets of Langerhans of patients with noninsulin-dependent diabetes mellitus.
Catalog No. NC0054993