Learn More
Bon Opus Biosciences Biotinylated Recombinant Human CD276/B7-H3 (C-Avi-His) 25ug
SDP
Supplier:  Bon Opus Biosciences BP111725UG
Encompass_Preferred
Biotinylated Recombinant Human CD276/B7-H3 (C-Avi-His) is produced by Human Cells. The target gene encoding leu29-pro245 is expressed with a C-Avi-His tag. Accession# Q5ZPR3-2. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1117
Catalog No. 50-217-2712