Learn More
Bon Opus Biosciences Biotinylated Recombinant Human CD22 (C-hFc) 25ug
SDP
Supplier:  Bon Opus Biosciences BP111525UG
Encompass_Preferred
Biotinylated Recombinant Human CD22 (C-hFc) is produced by Human Cells. The target gene encoding Asp20-Arg687 is expressed with a C-hFc, Avi tag. Accession# P20273-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1115
Catalog No. 50-217-2710