Learn More
Bioworld Biotin Hydrazide, 100 mg
Catalog No. 50255393
Encompass_Preferred
Biotin Hydrazide is a carbohydrate reactive biotinylation reagent. Biotin Hydrazide is used for labeling mouse monoclonal antibodies, unpaired cytosine residues in nucleic acids and it is also used fo coupling to glycoproteins through the carbohydrate by hydrazone formation.Carbohydrate reactivebiotin hydrazide is a a simplest hydrazide-activated biotinylation reagentMicroscopy studies on membranesAlternative names : Biotin-hydrazide,Hydrazide-activated biotinReactive groups Hydrazide, reacts with aldehydes at pH 4 to 6Click here to view chemical structures of Hydrazide-Biotin compounds. References: 1.) O'Shannessy, D.J., et al. (1984) Immun. Letters 8, 273-277. 2.) Heitzmann, H., et al. (1974) PNAS 71, 3537-3541. 3.) Lisanti, M.P., et al. (1989) J. Cell Biol. 109, 2117-2127.Appearance: White powder
Catalog No. 50-255-393 Supplier Bioworld Supplier No. 402000401
Biotin Hydrazide is a carbohydrate reactive biotinylation reagent. Biotin Hydrazide is used for labeling mouse monoclonal antibodies, unpaired cytosine residues in nucleic acids and it is also used fo coupling to glycoproteins through the carbohydrate by hydrazone formation.Carbohydrate reactivebiotin hydrazide is a a simplest hydrazide-activated biotinylation reagentMicroscopy studies on membranesAlternative names : Biotin-hydrazide,Hydrazide-activated biotinReactive groups Hydrazide, reacts with aldehydes at pH 4 to 6Click here to view chemical structures of Hydrazide-Biotin compounds. References: 1.) O'Shannessy, D.J., et al. (1984) Immun. Letters 8, 273-277. 2.) Heitzmann, H., et al. (1974) PNAS 71, 3537-3541. 3.) Lisanti, M.P., et al. (1989) J. Cell Biol. 109, 2117-2127.Appearance: White powder