Learn More
Apexbio Technology LLC Biotin Hydrazide, 1g. Cas: 66640-86-6 MFCD: MFCD00078532. Carbonyl-reactive biotinylation reagent
SDP
Supplier:  Apexbio Technology LLC A80071000
Encompass_Preferred
Biotin hydrazide is a biotinyl derivative that can be used as a probe for the determination of protein carbonylation, which is a component of several diseases. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4977