Learn More
Cayman Chemical BIotIn hydrazIde 1g
SDP
Catalog No. 502967592
Encompass_Preferred
A biotinyl derivative used to label surface functional groups, antibodies, lectins, sugars, nucleic acids, or molecules with free carboxylic or keto groups; probe for determining protein carbonylation
Catalog No. 50-296-7592 Supplier Cayman Chemical Supplier No. 184181 G
A biotinyl derivative used to label surface functional groups, antibodies, lectins, sugars, nucleic acids, or molecules with free carboxylic or keto groups; probe for determining protein carbonylation