Learn More
Cayman Chemical BIotIn hydrazIde 250mg
SDP
Catalog No. 502980118
Encompass_Preferred
A biotinyl derivative used to label surface functional groups, antibodies, lectins, sugars, nucleic acids, or molecules with free carboxylic or keto groups; probe for determining protein carbonylation
Catalog No. 50-298-0118 Supplier Cayman Chemical Supplier No. 18418250 MG
A biotinyl derivative used to label surface functional groups, antibodies, lectins, sugars, nucleic acids, or molecules with free carboxylic or keto groups; probe for determining protein carbonylation