Learn More
Cayman Chemical BIotIn hydrazIde 500mg
SDP
Catalog No. 502980448
Encompass_Preferred
A biotinyl derivative used to label surface functional groups, antibodies, lectins, sugars, nucleic acids, or molecules with free carboxylic or keto groups; probe for determining protein carbonylation
Catalog No. 50-298-0448 Supplier Cayman Chemical Supplier No. 18418500 MG
A biotinyl derivative used to label surface functional groups, antibodies, lectins, sugars, nucleic acids, or molecules with free carboxylic or keto groups; probe for determining protein carbonylation