Learn More
Medchemexpress LLC Biotin hydrazide | 66640-86-6 | MFCD00078532 | 258.34 g/mol | C10H18N4O2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10021510MM 1ML
Encompass_Preferred
Biotin hydrazide is a carbonyl-reactive biotinylation reagent used to label oxidized glycans, glycoproteins, and other carbonyl-containing biomolecules. It is supplied as a solid or as a 10 mM solution in DMSO for use in conjugation, detection, and enrichment workflows.
  • Reacts with carbonyl groups to form stable hydrazone linkages.
  • Supplied as solid or as a 10 mM solution in DMSO for convenience.
  • Enables biotin-based detection and affinity purification workflows.
  • Soluble in DMSO and moderately soluble in water, with solubility data provided.
  • Stable when stored under recommended temperatures for long-term use.
Catalog No. 50-002-15318