Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin hydrazide | 66640-86-6 | MFCD00078532 | 98.7% | 258.34 | C10H18N4O2S | 250 MG
SDP

Catalog No. 5000215321
Encompass_Preferred

Biotin hydrazide is a carbonyl-reactive biotinylation reagent used to label carbonyl-containing biomolecules such as glycoproteins and glycolipids for detection, enrichment, or affinity-capture applications.

  • Reacts selectively with carbonyl groups to introduce a biotin tag.
  • Suitable for biotinylation of glycoproteins and glycolipids.
  • High purity (98.7%) for consistent experimental results.
  • Soluble in DMSO (6.6 mg/mL) and water (2 mg/mL) with ultrasonic assistance.
  • Chemical formula C10H18N4O2S and molecular weight 258.34 g/mol.
  • Store powder at -20°C for long-term stability; 4°C for shorter-term storage.

Catalog No. 50-002-15321 Supplier Medchemexpress LLC Supplier No. HY100215250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin hydrazide is a carbonyl-reactive biotinylation reagent used to label carbonyl-containing biomolecules such as glycoproteins and glycolipids for detection, enrichment, or affinity-capture applications.

  • Reacts selectively with carbonyl groups to introduce a biotin tag.
  • Suitable for biotinylation of glycoproteins and glycolipids.
  • High purity (98.7%) for consistent experimental results.
  • Soluble in DMSO (6.6 mg/mL) and water (2 mg/mL) with ultrasonic assistance.
  • Chemical formula C10H18N4O2S and molecular weight 258.34 g/mol.
  • Store powder at -20°C for long-term stability; 4°C for shorter-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.