Learn More
Medchemexpress LLC Biotin hydrazide | 66640-86-6 | MFCD00078532 | 98.7% | 258.34 g/mol | C10H18N4O2S | 1 G
SDP
Catalog No. 5000215816
Encompass_Preferred
Biotin hydrazide is a carbonyl-reactive biotinylation reagent that forms hydrazone linkages with oxidized carbohydrates and other carbonyl groups. It is used to introduce a biotin tag for detection, affinity capture, and enrichment of labeled proteins and glycoconjugates in biochemical assays and proteomics workflows.
  • Reacts selectively with aldehydes and ketones after oxidation.
  • Enables streptavidin-based detection and affinity purification.
  • Suitable for labeling glycoproteins following periodate oxidation.
  • Supplied as a solid reagent for solution preparation and storage.
  • High purity for reproducible analytical and preparative work.
Catalog No. 50-002-15816 Supplier Medchemexpress LLC Supplier No. HY1002151G
Biotin hydrazide is a carbonyl-reactive biotinylation reagent that forms hydrazone linkages with oxidized carbohydrates and other carbonyl groups. It is used to introduce a biotin tag for detection, affinity capture, and enrichment of labeled proteins and glycoconjugates in biochemical assays and proteomics workflows.
  • Reacts selectively with aldehydes and ketones after oxidation.
  • Enables streptavidin-based detection and affinity purification.
  • Suitable for labeling glycoproteins following periodate oxidation.
  • Supplied as a solid reagent for solution preparation and storage.
  • High purity for reproducible analytical and preparative work.