Learn More
Medchemexpress LLC Biotin/KLH | 1 MG
SDP
Catalog No. 5000393047
Encompass_Preferred
Biotin/KLH is an antigen-adjuvant conjugate composed of Biotin and Keyhole Limpet Hemocyanin (KLH). This conjugate is designed to enhance the production of antigen-specific antibodies in vaccine models by conjugating the antigen with a protein adjuvant. It maintains protein folding, does not disrupt major epitopes, and can improve cross-presentation and the generation of antigen-specific T cells.
  • Enhances the production of antigen-specific antibodies in vaccine models.
  • Does not affect protein folding or disrupt major epitopes.
  • Can enhance cross-presentation and the generation of antigen-specific T cells.
Catalog No. 50-003-93047 Supplier Medchemexpress LLC Supplier No. HY1615531MG
Biotin/KLH is an antigen-adjuvant conjugate composed of Biotin and Keyhole Limpet Hemocyanin (KLH). This conjugate is designed to enhance the production of antigen-specific antibodies in vaccine models by conjugating the antigen with a protein adjuvant. It maintains protein folding, does not disrupt major epitopes, and can improve cross-presentation and the generation of antigen-specific T cells.
  • Enhances the production of antigen-specific antibodies in vaccine models.
  • Does not affect protein folding or disrupt major epitopes.
  • Can enhance cross-presentation and the generation of antigen-specific T cells.