Learn More
Medchemexpress LLC Biotin-labeled agatolimod sodium | 00-00-0 | 97.2% | 10MG
SDP
Catalog No. 5000258731
Encompass_Preferred
Biotin-labeled agatolimod sodium is a biotin-conjugated CpG oligodeoxynucleotide sodium salt that functions as a Toll-like receptor 9 (TLR9) agonist for immunology research. Supplied as a white to off-white solid with high reported purity, it is intended for TLR activation assays, biotin-based affinity capture, and detection workflows.
  • Biotin tag enables streptavidin-based capture and detection.
  • Acts as a TLR9 agonist for immune activation studies.
  • High purity suitable for in vitro assays.
  • Supplied as a sodium salt solid for stable handling.
  • Storage compatible with frozen, sealed, moisture-free conditions.
Catalog No. 50-002-58731 Supplier Medchemexpress LLC Supplier No. HY146244A10MG
Biotin-labeled agatolimod sodium is a biotin-conjugated CpG oligodeoxynucleotide sodium salt that functions as a Toll-like receptor 9 (TLR9) agonist for immunology research. Supplied as a white to off-white solid with high reported purity, it is intended for TLR activation assays, biotin-based affinity capture, and detection workflows.
  • Biotin tag enables streptavidin-based capture and detection.
  • Acts as a TLR9 agonist for immune activation studies.
  • High purity suitable for in vitro assays.
  • Supplied as a sodium salt solid for stable handling.
  • Storage compatible with frozen, sealed, moisture-free conditions.