Learn More
Medchemexpress LLC Biotin-labeled ODN 1826 sodium | 00-00-0 | 95.9% | 10MG
SDP
Catalog No. 5000258952
Encompass_Preferred
Biotin-labeled ODN 1826 sodium is a biotin-conjugated class B CpG oligodeoxynucleotide and a Toll-like receptor 9 (TLR9) agonist provided as a solid sodium salt for research use, useful for studying CpG uptake, localization, and TLR9 activation.
Catalog No. 50-002-58952 Supplier Medchemexpress LLC Supplier No. HY146245A10MG
Biotin-labeled ODN 1826 sodium is a biotin-conjugated class B CpG oligodeoxynucleotide and a Toll-like receptor 9 (TLR9) agonist provided as a solid sodium salt for research use, useful for studying CpG uptake, localization, and TLR9 activation.