Learn More
Invivogen ODN 2006 Biotin, 2 x 50 ug
SDP
Catalog No. NC0421503
Encompass
CpG ODNs are synthetic oligonucleotides that contain unmethylated CpG dinucleotides in particular sequence contexts (CpG motifs). These CpG motifs are present at a 20-fold greater frequency in bacterial DNA compared to mammalian DNA. They have been shown to induce a coordinated set of immune responses based on the activation of immune cells primarily involved in the recognition of these molecules.
Catalog No. NC0421503 Supplier Invivogen Supplier No. TLRL2006B
CpG ODNs are synthetic oligonucleotides that contain unmethylated CpG dinucleotides in particular sequence contexts (CpG motifs). These CpG motifs are present at a 20-fold greater frequency in bacterial DNA compared to mammalian DNA. They have been shown to induce a coordinated set of immune responses based on the activation of immune cells primarily involved in the recognition of these molecules.