Learn More
Medchemexpress LLC Biotin-labeled ODN 2395 sodium | 1254617-22-5 | 1 MG
SDP
Catalog No. 5000374842
Encompass_Preferred
Biotin-labeled ODN 2395 sodium is a C class oligodeoxynucleotide and a TLR9 agonist. It is used for research purposes to evaluate CpG ODN cellular uptake and localization using a biotin detection system and light microscopy.
  • Is a C class oligodeoxynucleotide
  • Acts as a TLR9 agonist
  • Facilitates evaluation of CpG ODN cellular uptake and localization
  • Designed for use with biotin detection systems and light microscopy
Catalog No. 50-003-74842 Supplier Medchemexpress LLC Supplier No. HY150743A1MG
Biotin-labeled ODN 2395 sodium is a C class oligodeoxynucleotide and a TLR9 agonist. It is used for research purposes to evaluate CpG ODN cellular uptake and localization using a biotin detection system and light microscopy.
  • Is a C class oligodeoxynucleotide
  • Acts as a TLR9 agonist
  • Facilitates evaluation of CpG ODN cellular uptake and localization
  • Designed for use with biotin detection systems and light microscopy