Learn More
Echelon Biosciences Research Labs Biotin Phosphatidylinositol 4-phosphate 10 ug
SDP
Supplier:  Echelon Biosciences Research Labs C04B610UG
Encompass
Biotin Phosphatidylinositol 4-phosphate is a water soluble analog of PI(4)P labeled with biotin at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4-phosphate (PI(4)P) is the biosynthetic precursor to PI(4,5)P2 and has an important roles in regulating sphingomyelin and glycosphingolipid metabolism and membrane trafficking at the exit of the Golgi complex. This item is non returnable
Catalog No. 50-720-055