Learn More
Echelon Biosciences Research Labs Biotin Phosphatidylinositol 5-phosphate 50 ug
SDP
Supplier:  Echelon Biosciences Research Labs C05B650UG
Encompass
Biotin Phosphatidylinositol 5-phosphate is a water soluble analog of PI(5)P labeled with biotin at the sn-1 position. This item is non returnable
Catalog No. 50-720-070