Learn More
Cayman Chemical BIotIn LC hydrazIde 100mg
SDP
Catalog No. 502985827
Encompass_Preferred
A reactive probe that is used to biotinylate carboxyl or vicinal hydroxyl groups on carbohydrates, glycoproteins, or other glycoconjugates; also used to link biotin to antibodies and ribonucleotides
Catalog No. 50-298-5827 Supplier Cayman Chemical Supplier No. 21497100 MG
A reactive probe that is used to biotinylate carboxyl or vicinal hydroxyl groups on carbohydrates, glycoproteins, or other glycoconjugates; also used to link biotin to antibodies and ribonucleotides