Learn More
Medchemexpress LLC Biotin LC hydrazide | 109276-34-8 | MFCD00077659 | 99.8% | 371.50 g/mol | C16H29N5O3S | 50 MG
SDP
Catalog No. 5000247943
Encompass_Preferred
Biotin LC hydrazide is a long-chain hydrazide reagent used to introduce a biotin tag onto periodate-oxidized glycans and glycoproteins. It labels aldehyde groups created by mild oxidation, delivering a biotin tag with an extended spacer arm that improves accessibility for detection and capture in downstream assays.
  • Reacts with periodate-oxidized glycoproteins and carbohydrate aldehydes.
  • Provides an extended spacer arm for improved streptavidin accessibility.
  • Suitable for biotinylation of proteins, glycans, and nucleic acids.
  • High purity suitable for analytical and preparative applications.
  • Provided as a solid in small, research-scale quantities.
Catalog No. 50-002-47943 Supplier Medchemexpress LLC Supplier No. HY10188550MG
Biotin LC hydrazide is a long-chain hydrazide reagent used to introduce a biotin tag onto periodate-oxidized glycans and glycoproteins. It labels aldehyde groups created by mild oxidation, delivering a biotin tag with an extended spacer arm that improves accessibility for detection and capture in downstream assays.
  • Reacts with periodate-oxidized glycoproteins and carbohydrate aldehydes.
  • Provides an extended spacer arm for improved streptavidin accessibility.
  • Suitable for biotinylation of proteins, glycans, and nucleic acids.
  • High purity suitable for analytical and preparative applications.
  • Provided as a solid in small, research-scale quantities.