Learn More
Cayman Chemical Biotin LC hydrazide, Purity: ≥98%, CAS: 109276-34-8, 250 mg
SDP
Catalog No. 502985828
Encompass_Preferred
A reactive probe that is used to biotinylate carboxyl or vicinal hydroxyl groups on carbohydrates, glycoproteins, or other glycoconjugates; also used to link biotin to antibodies and ribonucleotides
Catalog No. 50-298-5828 Supplier Cayman Chemical Supplier No. 21497250 MG
A reactive probe that is used to biotinylate carboxyl or vicinal hydroxyl groups on carbohydrates, glycoproteins, or other glycoconjugates; also used to link biotin to antibodies and ribonucleotides